[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-4-(hydroxymethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
PC6319-1G
APOSPC6319-1GEA
275
EUR
APOSPC6319-1G
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol 95%
[5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Formel:
C₆H₆ClF₃N₂O Molecular Weight: 214,58 g/mol Siedepunkt: 94…95 °C/0,8mmHg Schmelzpunkt: 94…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00141467 CAS-Nummer: 282523-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...