5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC6322-1G
PC6322-250MG
APOSPC6322-1GEA
0
EUR
APOSPC6322-1G
APOSPC6322-250MG
5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%
5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid
Formel:
C₁₄H₉ClF₃NO₃ Schmelzpunkt: 218…220 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038241 CAS-Nummer: 886761-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...