5-[(4-Chlorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%

Lieferant: Apollo Scientific

PC6337-1G
APOSPC6337-1GEA 355 EUR
APOSPC6337-1G
5-[(4-Chlorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde 97%
5-[(4-Chlorophenyl)sulphanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Formel: C₁₂H₈ClF₃N₂OS
Molecular Weight: 320,72 g/mol
Schmelzpunkt: 93…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00140822
CAS-Nummer: 321533-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR