2-Fluor-3-(trifluormethoxy)anilin 97%

Lieferant: Apollo Scientific
Achtung

PC6356-5G PC6356-1G PC6356-10G
APOSPC6356-5GEA 0 EUR
APOSPC6356-5G APOSPC6356-1G APOSPC6356-10G
2-Fluor-3-(trifluormethoxy)anilin 97%
2-Fluor-3-(trifluormethoxy)anilin
Formel: C₇H₅F₄NO
Lagertemperatur: Kühlschrank
MDL: MFCD11975313
CAS-Nummer: 1159512-64-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR