2-Fluoro-3-(trifluoromethoxy)benzylamine

Lieferant: Apollo Scientific
Gefahr

PC6357-1G PC6357-250MG PC6357-100MG
APOSPC6357-1GEA 0 EUR
APOSPC6357-1G APOSPC6357-250MG APOSPC6357-100MG
2-Fluoro-3-(trifluoromethoxy)benzylamine
2-Fluoro-3-(trifluoromethoxy)benzylamine
Formel: C₈H₇F₄NO
Lagertemperatur: Raumtemperatur
MDL: MFCD12026479
CAS-Nummer: 1159512-65-8
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR