3-Fluor-4-methoxybenzylamin 98%

Lieferant: Apollo Scientific
Gefahr

PC6364-5G PC6364-1G
APOSPC6364-5GEA 0 EUR
APOSPC6364-5G APOSPC6364-1G
3-Fluor-4-methoxybenzylamin 98%
3-Fluor-4-methoxybenzylamin
Formel: C₈H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD04116360
CAS-Nummer: 123652-95-9
UN: 2735
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR