3-Fluor-4-methoxybenzylamin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(3-Fluoro-4-methoxyphenyl)methanamine
PC6364-5G
APOSPC6364-5GEA
85.7
EUR
APOSPC6364-5G
3-Fluor-4-methoxybenzylamin 98%
3-Fluor-4-methoxybenzylamin
Formel:
C₈H₁₀FNO Molecular Weight: 155,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116360 CAS-Nummer: 123652-95-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...