4-(4-Fluorophenyl)-4-hydroxypiperidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 4-(4-Fluorophenyl)piperidin-4-ol hydrochloride

PC6366-1G
APOSPC6366-1GEA 190 EUR
APOSPC6366-1G
4-(4-Fluorophenyl)-4-hydroxypiperidine hydrochloride
4-(4-Fluorophenyl)-4-hydroxypiperidine hydrochloride
Formel: C₁₁H₁₄FNO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD06795784
CAS-Nummer: 3929-30-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR