3-[(3-Fluorophenyl)amino]propanoic acid

Lieferant: Apollo Scientific

Synonyme: 3-[(3-Fluorophenyl)amino]propionic acid, 3-((3-Fluorophenyl)amino)propanoic acid

PC6367-1G PC6367-5G
APOSPC6367-1GEA 125 EUR
APOSPC6367-1G APOSPC6367-5G
3-[(3-Fluorophenyl)amino]propanoic acid
3-[(3-Fluorophenyl)amino]propanoic acid
Formel: C₉H₁₀FNO₂
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784303
CAS-Nummer: 885275-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR