1-Methyl-5-(trifluoromethyl)-1H-indazol-3-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-1-methyl-5-(trifluoromethyl)-1H-indazole

PC6371-1G PC6371-250MG PC6371-5G
APOSPC6371-1GEA 174 EUR
APOSPC6371-1G APOSPC6371-250MG APOSPC6371-5G
1-Methyl-5-(trifluoromethyl)-1H-indazol-3-amine
1-Methyl-5-(trifluoromethyl)-1H-indazol-3-amine
Formel: C₉H₈F₃N₃
Molecular Weight: 215.18 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07777192
CAS-Nummer: 5685-69-8
UN: 2811
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR