2-Fluoro-DL-tyrosine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-(2-fluoro-4-hydroxyphenyl)propanoic acid, H-DL-Tyr(2-F)-OH

PC6380-1G PC6380-250MG
APOSPC6380-1GEA 835 EUR
APOSPC6380-1G APOSPC6380-250MG
2-Fluoro-DL-tyrosine 97%
2-Fluoro-DL-tyrosine
Formel: C₉H₁₀FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03005295
CAS-Nummer: 7656-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR