2-Fluoro-DL-tyrosine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-3-(2-fluoro-4-hydroxyphenyl)propanoic acid, H-DL-Tyr(2-F)-OH
PC6380-1G
PC6380-250MG
APOSPC6380-1GEA
835
EUR
APOSPC6380-1G
APOSPC6380-250MG
2-Fluoro-DL-tyrosine 97%
2-Fluoro-DL-tyrosine
Formel:
C₉H₁₀FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03005295 CAS-Nummer: 7656-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...