4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC6381-1G
APOSPC6381-1GEA 219 EUR
APOSPC6381-1G
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde
4-{[6-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde
Formel: C₁₃H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10703518
CAS-Nummer: 1086379-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR