(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Aminomethy-4-(4-fluorobenzyl)morpholine

PC6382-50G
APOSPC6382-50GEA 328 EUR
APOSPC6382-50G
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine
Formel: C₁₂H₁₇FN₂O
Molecular Weight: 224,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06658145
CAS-Nummer: 112914-13-3
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR