2-Fluoro-4-formylbenzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC6386-1G PC6386-5G PC6386-10G
APOSPC6386-1GEA 145 EUR
APOSPC6386-1G APOSPC6386-5G APOSPC6386-10G
2-Fluoro-4-formylbenzoic acid 97%
2-Fluoro-4-formylbenzoic acid
Formel: C₈H₅FO₃
Schmelzpunkt: 216 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12026481
CAS-Nummer: 604000-97-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR