4-Methoxy-2-(trifluormethoxy)benzylbromid 97%

Lieferant: Apollo Scientific

PC6389-10G PC6389-5G PC6389-1G
APOSPC6389-10GEA 405 EUR
APOSPC6389-10G APOSPC6389-5G APOSPC6389-1G
4-Methoxy-2-(trifluormethoxy)benzylbromid 97%
4-Methoxy-2-(trifluormethoxy)benzylbromid
Formel: C₉H₈BrF₃O₂
Molecular Weight: 285.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660289
CAS-Nummer: 886502-56-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR