4-Methoxy-2-(trifluormethoxy)benzylbromid 97%
Lieferant: Apollo Scientific
PC6389-10G
PC6389-5G
PC6389-1G
APOSPC6389-10GEA
405
EUR
APOSPC6389-10G
APOSPC6389-5G
APOSPC6389-1G
4-Methoxy-2-(trifluormethoxy)benzylbromid 97%
4-Methoxy-2-(trifluormethoxy)benzylbromid
Formel:
C₉H₈BrF₃O₂ Molecular Weight: 285.06 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660289 CAS-Nummer: 886502-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...