4-Methoxy-2-(trifluoromethoxy)benzoyl chloride 97%

Lieferant: Apollo Scientific

PC6391-5G PC6391-1G
APOSPC6391-5GEA 180 EUR
APOSPC6391-5G APOSPC6391-1G
4-Methoxy-2-(trifluoromethoxy)benzoyl chloride 97%
4-Methoxy-2-(trifluoromethoxy)benzoyl chloride
Formel: C₉H₆ClF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06660286
CAS-Nummer: 886502-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR