3-Fluor-4-(trifluormethoxy)benzonitril
Lieferant: Apollo Scientific
|
Achtung
|
PC6423-1G
PC6423-25G
PC6423-5G
APOSPC6423-1GEA
0
EUR
APOSPC6423-1G
APOSPC6423-25G
APOSPC6423-5G
3-Fluor-4-(trifluormethoxy)benzonitril
3-Fluor-4-(trifluormethoxy)benzonitril
Formel:
C₈H₃F₄NO Siedepunkt: 60 °C/ 5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660203 CAS-Nummer: 886498-94-8 UN: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...