5-(3-Chlorophenyl)-2-(trifluoromethyl)-3-furoic acid

Lieferant: Apollo Scientific

Synonyme: 5-(3-Chlorophenyl)-2-(trifluoromethyl)furan-3-carboxylic acid

PC6439-1G
APOSPC6439-1GEA 435 EUR
APOSPC6439-1G
5-(3-Chlorophenyl)-2-(trifluoromethyl)-3-furoic acid
5-(3-Chlorophenyl)-2-(trifluoromethyl)-3-furoic acid
Formel: C₁₂H₆ClF₃O₃
Molecular Weight: 290,63 g/mol
Schmelzpunkt: 164…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01313475
CAS-Nummer: 242812-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR