1,1,2-Trifluordecen 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC6459-25G
APOSPC6459-25GEA
248
EUR
APOSPC6459-25G
1,1,2-Trifluordecen 97%
1,1,2-Trifluordecen
Formel:
C₁₀H₁₇F₃ Siedepunkt: 91 °C/90mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156047 CAS-Nummer: 692-05-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...