1,1,1,2,2,3,4,5,5,5-Decafluor-3-methoxy-4-(trifluormethyl)pentan
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methoxyperfluoro(2-methylpentane)
PC6474-25G
APOSPC6474-25GEA
50.9
EUR
APOSPC6474-25G
1,1,1,2,2,3,4,5,5,5-Decafluor-3-methoxy-4-(trifluormethyl)pentan
1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane
Formel:
C₇H₃F₁₃O Molecular Weight: 350,08 g/mol Siedepunkt: 98 °C Dichte: 1,6 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD07784227 CAS-Nummer: 132182-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...