3-(2-Fluorophenoxy)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
PC6515-100MG
PC6515-250MG
APOSPC6515-100MGEA
0
EUR
APOSPC6515-100MG
APOSPC6515-250MG
3-(2-Fluorophenoxy)benzyl bromide
3-(2-Fluorophenoxy)benzyl bromide
Formel:
C₁₃H₁₀BrFO Dichte: 1,46 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00059914 CAS-Nummer: 242812-04-0 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...