4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Nitro-3-(trifluoromethyl)benzene-1-sulphonyl chloride
PC6567-1G
PC6567-5G
APOSPC6567-1GEA
102
EUR
APOSPC6567-1G
APOSPC6567-5G
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃ClF₃NO₄S Schmelzpunkt: 61…67 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094387 CAS-Nummer: 39234-83-8 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...