HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)

Lieferant: Apollo Scientific
Achtung

Synonyme: HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)

PC6587-100G PC6587-1KG PC6587-500G
APOSPC6587-100GEA 21.9 EUR
APOSPC6587-100G APOSPC6587-1KG APOSPC6587-500G
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)

Peptide coupling reagent.

Formel: C₁₁H₁₆F₆N₅OP
Molecular Weight: 379.2 g/mol
Schmelzpunkt: 200 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00075445
CAS-Nummer: 94790-37-1
EINECS: 423-020-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR