1H,1H-Perfluoro-3,6-dioxadecan-1-ol
Lieferant: Apollo Scientific
|
Achtung
|
PC6599-100G
PC6599-25G
PC6599-5G
PC6599-1G
APOSPC6599-100GEA
0
EUR
APOSPC6599-100G
APOSPC6599-25G
APOSPC6599-5G
APOSPC6599-1G
1H,1H-Perfluoro-3,6-dioxadecan-1-ol
1H,1H-Perfluoro-3,6-dioxadecan-1-ol
Formel:
C₈H₃F₁₅O₃ Siedepunkt: 150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094403 CAS-Nummer: 152914-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...