4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2,3,5,6-Tetrafluoro-4-(methoxymethyl)phenyl)methanol, 4-(Methoxymethyl)-2,3,5,6-tetrafluorobenzyl alcohol
PC6638-1G
APOSPC6638-1GEA
21.9
EUR
APOSPC6638-1G
4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol 97%
4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol
Formel:
C₉H₈F₄O₂ Molecular Weight: 224.16 g/mol Schmelzpunkt: 45…50 °C Flammpunkt: 107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08688060 CAS-Nummer: 83282-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...