2-Cyano-3-fluorophenylzinc iodide 0.5M in THF

Lieferant: Apollo Scientific

PC6661-50ML
APOSPC6661-50MLEA 452 EUR
APOSPC6661-50ML
2-Cyano-3-fluorophenylzinc iodide 0.5M in THF
2-Cyano-3-fluorophenylzinc iodide
Formel: C₇H₃FINZn
Molecular Weight: 312,417 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01319869
CAS-Nummer: 352525-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR