Tetraethylammoniumtetrafluoroborat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetrylammonium tetrafluoroborate

PC6667-25G
APOSPC6667-25GEA 49.3 EUR
APOSPC6667-25G
Tetraethylammoniumtetrafluoroborat 97%
Tetraethylammoniumtetrafluoroborat

Useful electrolyte for electro-organic synthesis, e.g. thehydroxylation by anodic oxidation of aromatic ketones, acids and esters in trifluoroacetic acid.

Formel: C₈H₂₀BF₄N
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011827
CAS-Nummer: 429-06-1
EINECS: 207-055-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR