4-(3-Fluorphenoxy)piperidin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

PC6669-1G PC6669-250MG
APOSPC6669-1GEA 105 EUR
APOSPC6669-1G APOSPC6669-250MG
4-(3-Fluorphenoxy)piperidin Hydrochlorid
4-(3-Fluorphenoxy)piperidin Hydrochlorid
Formel: C₁₁H₁₅ClFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD08445760
CAS-Nummer: 3202-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR