4-(3-Fluorphenoxy)piperidin Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
PC6669-1G
PC6669-250MG
APOSPC6669-1GEA
105
EUR
APOSPC6669-1G
APOSPC6669-250MG
4-(3-Fluorphenoxy)piperidin Hydrochlorid
4-(3-Fluorphenoxy)piperidin Hydrochlorid
Formel:
C₁₁H₁₅ClFNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445760 CAS-Nummer: 3202-36-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...