2-Amino-5-brom-3-(trifluormethyl)pyridin
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-3-(trifluoromethyl)pyridin-2-amine
PC6673-5G
APOSPC6673-5GEA
98.6
EUR
APOSPC6673-5G
2-Amino-5-brom-3-(trifluormethyl)pyridin
2-Amino-5-bromo-3-(trifluoromethyl)pyridine
Formel:
C₆H₄BrF₃N₂ Molecular Weight: 241,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10758088 CAS-Nummer: 79456-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...