3-Brom-4-fluorbiphenyl 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-4-fluoro-1,1'-biphenyl

PC6677-1G
APOSPC6677-1GEA 29.2 EUR
APOSPC6677-1G
3-Brom-4-fluorbiphenyl 97%
3-Brom-4-fluorbiphenyl
Formel: C₁₂H₈BrF
Molecular Weight: 251,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01571091
CAS-Nummer: 306935-88-6
UN: 3151
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR