Perfluoro(4,4'-diphenoxybiphenyl) 90%
Lieferant: Apollo Scientific
|
Achtung
|
PC6682-1G
PC6682-250MG
APOSPC6682-1GEA
458
EUR
APOSPC6682-1G
APOSPC6682-250MG
Perfluoro(4,4'-diphenoxybiphenyl) 90%
Perfluoro(4,4'-diphenoxybiphenyl)
Formel:
C₂₄F₁₈O₂ Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09763620 CAS-Nummer: 14055-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...