1,5-Difluoro-2,4-dimethoxybenzene 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,5-Difluoro-2,4-dimethoxybenzene, 4,6-Difluororesorcinol dimethyl ether
PC6689-1G
APOSPC6689-1GEA
456
EUR
APOSPC6689-1G
1,5-Difluoro-2,4-dimethoxybenzene 97%
1,5-Difluoro-2,4-dimethoxybenzene
Formel:
C₈H₈F₂O₂ Siedepunkt: 55 °C (50 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193047 CAS-Nummer: 79069-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...