2-Fluor-9-fluorenon 98%

Lieferant: Apollo Scientific
Achtung

PC6694-1G
APOSPC6694-1GEA 161 EUR
APOSPC6694-1G
2-Fluor-9-fluorenon 98%
2-Fluor-9-fluorenon
Formel: C₁₃H₇FO
Molecular Weight: 198.2 g/mol
Schmelzpunkt: 115…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001150
CAS-Nummer: 343-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR