1-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-(2-Chloro-6-fluorobenzyloxy)acetophenone

PC6706-1G
APOSPC6706-1GEA 50.9 EUR
APOSPC6706-1G
1-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone 97%
1-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1-ethanone
Formel: C₁₅H₁₂ClFO₂
Molecular Weight: 278,71 g/mol
Schmelzpunkt: 92…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179827
CAS-Nummer: 306934-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR