5-Fluoro-2-(trifluoromethyl)phenylacetonitrile 98%

Lieferant: Apollo Scientific
Achtung

PC6708-1G
APOSPC6708-1GEA 21.9 EUR
APOSPC6708-1G
5-Fluoro-2-(trifluoromethyl)phenylacetonitrile 98%
5-Fluoro-2-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₅F₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD06247720
CAS-Nummer: 242812-09-5
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR