5,6,7,8-Tetrahydro-2-thio-4-(trifluoromethyl)quinoline-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Cyano-5,6,7,8-tetrahydro-2-thio-4-(trifluoromethyl)quinoline

PC6727-250MG
APOSPC6727-250MGEA 105 EUR
APOSPC6727-250MG
5,6,7,8-Tetrahydro-2-thio-4-(trifluoromethyl)quinoline-3-carbonitrile
5,6,7,8-Tetrahydro-2-thio-4-(trifluoromethyl)quinoline-3-carbonitrile
Formel: C₁₁H₉F₃N₂S
Molecular Weight: 258,263 g/mol
Schmelzpunkt: 216…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02180672
CAS-Nummer: 306935-92-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR