Tetrafluor-p-benzochinon 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil

PC6740-1G
APOSPC6740-1GEA 36.4 EUR
APOSPC6740-1G
Tetrafluor-p-benzochinon 97%
Tetrafluor-p-benzochinon

High potential quinone; undergoes Diels-Alder cyclo-additions; complexes with a wide range of electron donors

Formel: C₆F₄O₂
Molecular Weight: 180.06 g/mol
Siedepunkt: 133
Schmelzpunkt: 183…186 °C (Subl.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001592
CAS-Nummer: 527-21-9
EINECS: 208-411-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR