4H-Tetrafluorophenyl methacrylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3,5,6-Tetrafluorophenyl methacrylate, 2,3,5,6-Tetrafluorophenyl 2-methylprop-2-enoate

PC6744-25G PC6744-5G
APOSPC6744-25GEA 379 EUR
APOSPC6744-25G APOSPC6744-5G
4H-Tetrafluorophenyl methacrylate 97%
4H-Tetrafluorophenyl methacrylate
Formel: C₁₀H₆F₄O₂
Molecular Weight: 234.147 g/mol
Dichte: 1.3 g/cm³ (25 °C)
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00129959
CAS-Nummer: 101156-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR