2-(1,1,2,2-Tetrafluorethoxy)nitrobenzol 98%
Lieferant: Apollo Scientific
PC6774-25G
APOSPC6774-25GEA
379
EUR
APOSPC6774-25G
2-(1,1,2,2-Tetrafluorethoxy)nitrobenzol 98%
2-(1,1,2,2-Tetrafluorethoxy)nitrobenzol
Formel:
C₈H₅F₄NO₃ Siedepunkt: 66…68 °C/1mmHg Dichte: 1,48 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
CAS-Nummer:
28202-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...