3-(1,1,2,2-Tetrafluorethoxy)phenol 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC6777G-5G
APOSPC6777G-5GEA
42.2
EUR
APOSPC6777G-5G
3-(1,1,2,2-Tetrafluorethoxy)phenol 97%
3-(1,1,2,2-Tetrafluorethoxy)phenol
Formel:
C₈H₆F₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153276 CAS-Nummer: 53997-99-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...