Heptafluorobutyryl bromide 97%

Lieferant: Apollo Scientific
Gefahr

PC6791-1G
APOSPC6791-1GEA 46.4 EUR
APOSPC6791-1G
Heptafluorobutyryl bromide 97%
Heptafluorobutyryl bromide
Formel: C₄BrF₇O
Siedepunkt: 52…54 °C (736 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00155814
CAS-Nummer: 375-13-3
UN: 1760
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR