4-(Trifluoromethoxy)benzene-1,3-diamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1,3-Diamino-4-(trifluoromethoxy)benzene

PC6792-1G
APOSPC6792-1GEA 58 EUR
APOSPC6792-1G
4-(Trifluoromethoxy)benzene-1,3-diamine
4-(Trifluoromethoxy)benzene-1,3-diamine
Formel: C₇H₇F₃N₂O
Molecular Weight: 192,138 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972990
CAS-Nummer: 873055-90-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR