5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Amino-4-carboxy-1H-pyrazol-1-yl)-4-fluorobenzene, 5-Amino-4-carboxy-1-(4-fluorophenyl)-1H-pyrazole

PC6796-250MG
APOSPC6796-250MGEA 21.9 EUR
APOSPC6796-250MG
5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylic acid
5-Amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylic acid
Formel: C₁₀H₈FN₃O₂
Molecular Weight: 221,188 g/mol
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01569434
CAS-Nummer: 187949-90-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR