Bis(perfluorophenyl)bromophosphine

Lieferant: Apollo Scientific
Achtung

Synonyme: Bis(pentafluorophenyl)phosphinous bromide

PC6803-1G
APOSPC6803-1GEA 408 EUR
APOSPC6803-1G
Bis(perfluorophenyl)bromophosphine
Bis(perfluorophenyl)bromophosphine
Formel: C₁₂BrF₁₀P
Molecular Weight: 444,99 g/mol
Siedepunkt: 112…114 °C (0,5 mm Hg)
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09763603
CAS-Nummer: 13648-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR