4-Bromo-2-(difluoromethyl)-1-fluorobenzene 97%

Lieferant: Apollo Scientific

PC6822-25G
APOSPC6822-25GEA 270 EUR
APOSPC6822-25G
4-Bromo-2-(difluoromethyl)-1-fluorobenzene 97%
4-Bromo-2-(difluoromethyl)-1-fluorobenzene
Formel: C₇H₄BrF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06411633
CAS-Nummer: 445303-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR