2-Bromo-4-fluorophenacyl bromide

Lieferant: Apollo Scientific
Gefahr

PC6831-1G PC6831-5G
APOSPC6831-1GEA 0 EUR
APOSPC6831-1G APOSPC6831-5G
2-Bromo-4-fluorophenacyl bromide
2-Bromo-4-fluorophenacyl bromide
Formel: C₈H₅Br₂FO
Lagertemperatur: Raumtemperatur
MDL: MFCD11847026
CAS-Nummer: 594810-90-9
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR