4-(Trifluormethoxy)benzolsulfonamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluormethoxy)benzolsulfonsäureamid
PC6842-1G
PC6842-5G
APOSPC6842-1GEA
14.7
EUR
APOSPC6842-1G
APOSPC6842-5G
4-(Trifluormethoxy)benzolsulfonamid
4-(Trifluormethoxy)benzolsulfonamid
Formel:
C₇H₆F₃NO₃S Molecular Weight: 241.19 g/mol Schmelzpunkt: 146…148 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798381 CAS-Nummer: 1513-45-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...